Assay Details
Gene Symbol: PLA2G4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252598.2 | XP_008250820.1 | 1277 | 87 | ||
XM_017339938.1 | XP_017195427.1 | 413 | 87 |

More Information
Set Membership: |
![]() ![]() |
