Assay Details
Gene Symbol: TASOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248603.2 | XP_008246825.1 | 809 | 110 | ||
XM_008248604.2 | XP_008246826.1 | 727 | 110 | |||
XM_008248605.2 | XP_008246827.1 | 1604 | 110 | |||
XM_008248608.2 | XP_008246830.1 | 1288 | 110 |

More Information
Set Membership: |
![]() ![]() |
