Assay Details
Gene Symbol: DNAI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008265088.2 | XP_008263310.1 | 1 - 2 | 93 | 83 | |
XM_017346207.1 | XP_017201696.1 | 1 - 2 | 93 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
