Assay Details
Gene Symbol: MAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719525.3 | XP_002719571.1 | 3 - 4 | 507 | 59 | |
XM_002719526.3 | XP_002719572.1 | 4 - 5 | 535 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
