Assay Details
Gene Symbol: RNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262860.2 | XP_008261082.1 | 2 - 3 | 327 | 63 | |
XM_017345090.1 | XP_017200579.1 | 2 - 3 | 330 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
