Assay Details
Gene Symbol: NDUFAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714055.2 | XP_002714101.1 | 3 - 4 | 348 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NDUFAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714055.2 | XP_002714101.1 | 3 - 4 | 348 | 135 |
Set Membership: |
![]() ![]() ![]() ![]() |