Assay Details
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017345966.1 | XP_017201455.1 | 2 - 3 | 406 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017345966.1 | XP_017201455.1 | 2 - 3 | 406 | 76 |
Set Membership: |
![]() ![]() ![]() |