Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017340645.1 | XP_017196134.1 | 1 - 2 | 108 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017340645.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
