Assay Details
Gene Symbol: GRPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710299.3 | XP_002710345.1 | 3 - 4 | 489 | 99 | |
XM_017341179.1 | XP_017196668.1 | 4 - 5 | 475 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
