Assay Details
Gene Symbol: ATF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008268442.2 | XP_008266664.1 | 1 - 2 | 150 | 138 | |
XM_017347798.1 | XP_017203287.1 | 1 - 2 | 151 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008268442.2, XM_017347798.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
