Assay Details
Gene Symbol: TMEM207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017346987.1 | XP_017202476.1 | 1 - 2 | 62 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017346987.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
