Assay Details
Gene Symbol: LOC100350827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262483.2 | XP_008260705.1 | 12 - 13 | 1636 | 69 | |
XM_008262484.2 | XP_008260706.1 | 10 - 11 | 1530 | 69 | ||
XM_017344959.1 | XP_017200448.1 | 13 - 14 | 1740 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
