Assay Details
Gene Symbol: LOC100350827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262483.2 | XP_008260705.1 | 5 - 6 | 624 | 104 | |
XM_008262484.2 | XP_008260706.1 | 3 - 4 | 518 | 104 | ||
XM_017344959.1 | XP_017200448.1 | 6 - 7 | 728 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
