Assay Details
Gene Symbol: LOC100350827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262483.2 | XP_008260705.1 | 6 - 7 | 819 | 74 | |
XM_008262484.2 | XP_008260706.1 | 4 - 5 | 713 | 74 | ||
XM_017344959.1 | XP_017200448.1 | 7 - 8 | 923 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
