Assay Details
Gene Symbol: TDGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008260522.2 | XP_008258744.1 | 6 - 7 | 1048 | 89 | |
XM_017343777.1 | XP_017199266.1 | 5 - 6 | 646 | 89 | ||
XM_017343778.1 | XP_017199267.1 | 6 - 7 | 683 | 89 | ||
XM_017343779.1 | XP_017199268.1 | 6 - 7 | 739 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
