Assay Details
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711627.3 | XP_002711673.2 | 7 - 8 | 790 | 60 | |
XM_008257488.2 | XP_008255710.1 | 6 - 7 | 885 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
