Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264256.2 | XP_008262478.1 | 12 - 13 | 1735 | 73 | |
XM_008264257.2 | XP_008262479.1 | 12 - 13 | 1687 | 73 | ||
XM_008264258.2 | XP_008262480.1 | 11 - 12 | 1585 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
