Assay Details
Gene Symbol: BCL2L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008268994.2 | XP_008267216.1 | 1 | 342 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008268994.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
