Assay Details
Gene Symbol: TENT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264592.2 | XP_008262814.1 | 2 | 5544 | 77 | |
XM_017345948.1 | XP_017201437.1 | 3 | 5716 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
