Assay Details
Gene Symbol: ATP5F1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717326.3 | XP_002717372.1 | 7 - 8 | 860 | 105 | |
XM_008268067.2 | XP_008266289.1 | 7 - 8 | 860 | 105 | ||
XM_008268068.2 | XP_008266290.1 | 7 - 8 | 860 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
