Assay Details
Gene Symbol: MRPL55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252677.2 | XP_008250899.1 | 1 | 93 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008252677.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
