Assay Details
Gene Symbol: LOC100345427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714462.3 | XP_002714508.2 | 6 - 7 | 1064 | 81 | |
XM_017345162.1 | XP_017200651.1 | 6 - 7 | 1066 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
