Assay Details
Gene Symbol: PRXL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720973.3 | XP_002721019.1 | 766 | 80 | ||
XM_008274310.2 | XP_008272532.1 | 666 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
