Assay Details
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259540.2 | XP_008257762.2 | 2 - 3 | 1300 | 94 | |
XM_017343338.1 | XP_017198827.1 | 2 - 3 | 1311 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
