Assay Details
Gene Symbol: CMPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721135.3 | XP_002721181.1 | 1070 | 80 | ||
XM_017350631.1 | XP_017206120.1 | 1076 | 80 |

More Information
Set Membership: |
![]() ![]() |
