Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017347739.1 | XP_017203228.1 | 11 - 12 | 1221 | 82 | |
XM_017347740.1 | XP_017203229.1 | 10 - 11 | 1152 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
