Assay Details
Gene Symbol: PCBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710218.3 | XP_002710264.1 | 1 - 2 | 331 | 110 | |
XM_017341067.1 | XP_017196556.1 | 1 - 2 | 444 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
