Assay Details
Gene Symbol: GPRC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712647.3 | XP_002712693.1 | 1 - 2 | 660 | 62 | |
XM_008259607.2 | XP_008257829.1 | 1 - 2 | 640 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712647.3, XM_008259607.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
