Assay Details
Gene Symbol: SMUG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008256556.2 | XP_008254778.1 | 2 - 3 | 1188 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008256556.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
