Assay Details
Gene Symbol: LOC100351437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017348165.1 | XP_017203654.1 | 13 - 14 | 2173 | 79 | |
XM_017348166.1 | XP_017203655.1 | 13 - 14 | 2115 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
