Assay Details
Gene Symbol: LOC100351437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017348165.1 | XP_017203654.1 | 14 - 15 | 2331 | 81 | |
XM_017348166.1 | XP_017203655.1 | 14 - 15 | 2273 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
