Assay Details
Gene Symbol: CKS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264365.2 | XP_008262587.2 | 1 - 2 | 154 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008264365.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
