Assay Details
Gene Symbol: MOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723758.3 | XP_002723804.1 | 571 | 101 | ||
XM_017339850.1 | XP_017195339.1 | 401 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
