Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709268.3 | XP_002709314.2 | 7 | 2713 | 102 | |
XM_017350642.1 | XP_017206131.1 | 7 | 2554 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
