Assay Details
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720502.3 | XP_002720548.1 | 795 | 63 | ||
XM_008273376.2 | XP_008271598.1 | 582 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
