Assay Details
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709431.3 | XP_002709477.1 | 5 - 6 | 414 | 84 | |
XM_008248769.2 | XP_008246991.1 | 5 - 6 | 422 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
