Assay Details
Gene Symbol: DUSP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709541.3 | XP_002709587.1 | 1 | 161 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002709541.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
