Assay Details
Gene Symbol: ACYP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008254402.2 | XP_008252624.1 | 1 - 2 | 416 | 66 | |
XM_008254403.2 | XP_008252625.1 | 1 - 2 | 433 | 66 | ||
XM_017340779.1 | XP_017196268.1 | 1 - 2 | 444 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
