Assay Details
Gene Symbol: LOC100349431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008269433.2 | XP_008267655.1 | 5 - 6 | 1143 | 76 | |
XM_008269434.2 | XP_008267656.1 | 4 - 5 | 1035 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
