Assay Details
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716437.3 | XP_002716483.1 | 3 - 4 | 1074 | 94 | |
XM_008266218.2 | XP_008264440.2 | 2 - 3 | 874 | 94 | ||
XM_008266219.2 | XP_008264441.1 | 3 - 4 | 1133 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
