Assay Details
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713020.3 | XP_002713066.1 | 10 - 11 | 1733 | 69 | |
XM_008260166.2 | XP_008258388.1 | 8 - 9 | 851 | 69 | ||
XM_017343581.1 | XP_017199070.1 | 10 - 11 | 1754 | 69 | ||
XM_017343582.1 | XP_017199071.1 | 10 - 11 | 1572 | 69 | ||
XM_017343583.1 | XP_017199072.1 | 10 - 11 | 1751 | 69 | ||
XM_017343584.1 | XP_017199073.1 | 9 - 10 | 982 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
