Assay Details
Gene Symbol: USF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716690.3 | XP_002716736.1 | 2 - 3 | 305 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002716690.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
