Assay Details
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714411.3 | XP_002714457.1 | 1 - 2 | 549 | 60 | |
XM_017345134.1 | XP_017200623.1 | 1 - 2 | 562 | 60 | ||
XM_017345135.1 | XP_017200624.1 | 1 - 2 | 556 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
