Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264899.2 | XP_008263121.1 | 1 - 2 | 305 | 74 | |
XM_008264904.2 | XP_008263126.1 | 1 - 2 | 304 | 74 | ||
XM_008264905.2 | XP_008263127.1 | 1 - 2 | 306 | 74 | ||
XM_008264907.2 | XP_008263129.1 | 1 - 2 | 293 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008264899.2, XM_008264904.2, XM_008264905.2, XM_008264907.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
