Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264899.2 | XP_008263121.1 | 3 - 4 | 3398 | 67 | |
XM_008264900.2 | XP_008263122.1 | 4 - 5 | 3996 | 67 | ||
XM_008264901.2 | XP_008263123.1 | 4 - 5 | 4000 | 67 | ||
XM_008264902.2 | XP_008263124.1 | 3 - 4 | 3394 | 67 | ||
XM_008264904.2 | XP_008263126.1 | 3 - 4 | 3397 | 67 | ||
XM_008264905.2 | XP_008263127.1 | 3 - 4 | 3399 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
