Assay Details
Gene Symbol: TNFAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718932.3 | XP_002718978.1 | 2 - 3 | 429 | 74 | |
XM_017348957.1 | XP_017204446.1 | 3 - 4 | 1410 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
