Assay Details
Gene Symbol: MACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713706.3 | XP_002713752.2 | 4 - 5 | 2542 | 102 | |
XM_008261503.2 | XP_008259725.1 | 4 - 5 | 2461 | 102 | ||
XM_008261504.2 | XP_008259726.1 | 5 - 6 | 2484 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
