Assay Details
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249776.2 | XP_008247998.1 | 3 - 4 | 544 | 73 | |
XM_008249777.2 | XP_008247999.1 | 2 - 3 | 549 | 73 | ||
XM_008249778.2 | XP_008248000.1 | 2 - 3 | 448 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
