Assay Details
Gene Symbol: CCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017349037.1 | XP_017204526.1 | 2 - 3 | 375 | 69 | |
GenBank mRNA | KF527431.1 | - | 1 - 2 | 76 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
