Assay Details
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274287.2 | XP_008272509.1 | 1887 | 76 | ||
XM_008274288.2 | XP_008272510.1 | 2451 | 76 | |||
XM_008274289.2 | XP_008272511.1 | 1551 | 76 | |||
XM_008274290.2 | XP_008272512.1 | 2691 | 76 | |||
XM_008274291.2 | XP_008272513.1 | 1962 | 76 | |||
XM_008274292.2 | XP_008272514.1 | 2061 | 76 | |||
XM_008274293.2 | XP_008272515.2 | 741 | 76 | |||
XM_017350515.1 | XP_017206004.1 | 1491 | 76 |

More Information
Set Membership: |
![]() ![]() |
